1-Naphthoxyacetic acid, 98+%
1-Naphthoxyacetic acid, 98+%
1-Naphthoxyacetic acid, 98+%
Thermo Scientific Chemicals

1-Naphthoxyacetic acid, 98+%

CAS: 2976-75-2 | C12H10O3 | 202.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01036.14
also known as L01036-14
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS2976-75-2
IUPAC Name2-(naphthalen-1-yloxy)acetic acid
Molecular FormulaC12H10O3
InChI KeyGHRYSOFWKRRLMI-UHFFFAOYSA-N
SMILESOC(=O)COC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown or pink
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0% [UK sourced material]
Assay (Silylated GC)≥98.0% [UK sourced material]
FormCrystals or powder or crystalline powder or fused solid
Assay from Supplier's CofA≥98.0% (H-NMR) [US sourced material]
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only