2-(8-Bromooctyloxy)tetrahydropyran, tech. 90%
2-(8-Bromooctyloxy)tetrahydropyran, tech. 90%
2-(8-Bromooctyloxy)tetrahydropyran, tech. 90%
2-(8-Bromooctyloxy)tetrahydropyran, tech. 90%
Thermo Scientific Chemicals

2-(8-Bromooctyloxy)tetrahydropyran, tech. 90%

CAS: 50816-20-1 | C13H25BrO2 | 293.245 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01081.14
also known as L01081-14
Price (USD)
193.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
193.00
Each
Add to cart
Chemical Identifiers
CAS50816-20-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4730-1.4810 @ 20?C
Assay (GC)≥88.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only