3-Methoxyphenoxyacetic acid, 97+%
3-Methoxyphenoxyacetic acid, 97+%
3-Methoxyphenoxyacetic acid, 97+%
Thermo Scientific Chemicals

3-Methoxyphenoxyacetic acid, 97+%

CAS: 2088-24-6 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01100.06
also known as L01100-06
Price (USD)
40.65
Online exclusive
44.70
Save 4.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Online exclusive
44.70
Save 4.05
Each
Chemical Identifiers
CAS2088-24-6
IUPAC Name2-(3-methoxyphenoxy)acetic acid
Molecular FormulaC9H10O4
InChI KeyAHDPQRIYMMZJTF-UHFFFAOYSA-N
SMILESCOC1=CC=CC(OCC(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Assay (Silylated GC)≥97.0%
Melting Point (clear melt)113.0-122.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only