4-Isopropylphenoxyacetic acid, 98+%
4-Isopropylphenoxyacetic acid, 98+%
4-Isopropylphenoxyacetic acid, 98+%
Thermo Scientific Chemicals

4-Isopropylphenoxyacetic acid, 98+%

CAS: 1643-16-9 | C11H14O3 | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01101.06
also known as L01101-06
Price (USD)
46.65
Online Exclusive
51.20
Save 4.55 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
51.20
Save 4.55 (9%)
Each
Chemical Identifiers
CAS1643-16-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only