5-Benzyloxyindole-3-carboxaldehyde, 98%
5-Benzyloxyindole-3-carboxaldehyde, 98%
5-Benzyloxyindole-3-carboxaldehyde, 98%
Thermo Scientific Chemicals

5-Benzyloxyindole-3-carboxaldehyde, 98%

CAS: 6953-22-6 | C16H13NO2 | 251.285 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L01117.06
also known as L01117-06
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
204.65
Online Exclusive
227.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS6953-22-6
IUPAC Name5-(benzyloxy)-1H-indole-3-carbaldehyde
Molecular FormulaC16H13NO2
InChI KeyDJGNUBADRQIDNQ-UHFFFAOYSA-N
SMILESO=CC1=CNC2=CC=C(OCC3=CC=CC=C3)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
Appearance (Color)Pale yellow to Dark Yellow or Pale Cream to Brown
Free acid (titration)<1.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only