Thiocarbohydrazide, 97%
Thiocarbohydrazide, 97%
Thiocarbohydrazide, 97%
Thermo Scientific Chemicals

Thiocarbohydrazide, 97%

CAS: 2231-57-4 | CH6N4S | 106.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L01205.06
also known as L01205-06
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.50
Save 6.85 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.50
Save 6.85 (14%)
Each
Chemical Identifiers
CAS2231-57-4
IUPAC Name1,3-diaminothiourea
Molecular FormulaCH6N4S
InChI KeyLJTFFORYSFGNCT-UHFFFAOYSA-N
SMILESNNC(=S)NN
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point159-180?C (dec)
Assay (unspecified)≥95.0%
Appearance (Color)White to pale cream to pale brown or grey
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

Solubility
Very slightly soluble (0.5g/100g, 25°C). Insoluble in ether.

Notes
Light Sensitive. Incompatible with strong acids, Strong oxidizing agents. Avoid heat.
RUO – Research Use Only

General References:

  1. Gary R. Burns. Metal complexes of thiocarbohydrazide.Inorg. Chem.1968, 7 (2), 277-283.
  2. L. F. Audrieth; Earle S. Scott; Perry S. Kippur. Hydrazine derivatives of the carbonic and thiocarbonic acids. I. The preparation and properties of thiocarbohydrazide.J. Org. Chem.1954, 19 (5), 733-741.
  3. For a review of the chemistry of carbohydrazide and thiocarbohydrazide, see: Chem. Rev., 70, 111 (1970).