2-Amino-1,3,4-thiadiazole, 98+%
2-Amino-1,3,4-thiadiazole, 98+%
2-Amino-1,3,4-thiadiazole, 98+%
Thermo Scientific Chemicals

2-Amino-1,3,4-thiadiazole, 98+%

CAS: 4005-51-0 | C2H3N3S | 101.127 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01244.06
also known as L01244-06
Price (USD)
44.70
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
44.70
Each
Chemical Identifiers
CAS4005-51-0
IUPAC Name1,3,4-thiadiazol-2-amine
Molecular FormulaC2H3N3S
InChI KeyQUKGLNCXGVWCJX-UHFFFAOYSA-N
SMILESNC1=NN=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey or cream
FormCrystalline powder
Assay (HPLC)>98.0%
2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.

Solubility
It is soluble in water, hot methanol- very faint turbidity.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. N Terzioglu.; A Gürsoy. Synthesis and anticancer evaluation of some new hydrazone derivatives of 2, 6-dimethylimidazo [2, 1-b][1, 3, 4] thiadiazole-5-carbohydrazide. European journal of medicinal chemistry. 200338(7-8) , 781-786.
  2. JA Nelson.; LM Rose.; LL Bennett. Mechanism of action of 2-amino-1, 3, 4-thiadiazole (NSC 4728). Cancer research. 197737(1) , 182-186.