3-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%
3-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%
3-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%
3-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%
Thermo Scientific Chemicals

3-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%

CAS: 21211-22-3 | C9H5ClO2S | 212.647 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L01257.03
also known as L01257-03
Price (USD)
36.65
Online Exclusive
40.40
Save 3.75 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
40.40
Save 3.75 (9%)
Each
Add to cart
Specifications
Beilstein1425602
CAS21211-22-3
Chemical Name or Material3-Chlorobenzo[b]thiophene-2-carboxylic acid
EINECS Number000-000-0
Formula Weight212.65
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only