N-(3-Pyridyl)thiourea, 98+%
N-(3-Pyridyl)thiourea, 98+%
N-(3-Pyridyl)thiourea, 98+%
Thermo Scientific Chemicals

N-(3-Pyridyl)thiourea, 98+%

CAS: 30162-37-9 | C6H7N3S | 153.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L01284.03
also known as L01284-03
Price (USD)
69.65
Online exclusive
76.90
Save 7.25
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
69.65
Online exclusive
76.90
Save 7.25
Each
Chemical Identifiers
CAS30162-37-9
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (HPLC)>98.0%
Appearance (Color)White to pale yellow, pale brown or pale grey
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only