2,5-Dimethylbenzyl chloride, 98%
2,5-Dimethylbenzyl chloride, 98%
2,5-Dimethylbenzyl chloride, 98%
Thermo Scientific Chemicals

2,5-Dimethylbenzyl chloride, 98%

CAS: 824-45-3 | C9H11Cl | 154.637 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01303.14
also known as L01303-14
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS824-45-3
IUPAC Name2-(chloromethyl)-1,4-dimethylbenzene
Molecular FormulaC9H11Cl
InChI KeyPECXPZGFZFGDRD-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C(CCl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5355-1.5400 @ 20?C
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only