(n-Hexadecyl)tri-n-butylphosphonium bromide, 98+%
(n-Hexadecyl)tri-n-butylphosphonium bromide, 98+%
(n-Hexadecyl)tri-n-butylphosphonium bromide, 98+%
Thermo Scientific Chemicals

(n-Hexadecyl)tri-n-butylphosphonium bromide, 98+%

CAS: 14937-45-2 | C28H60BrP | 507.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L01335.22
also known as L01335-22
Price (USD)
298.65
Special offer
Online exclusive
Ends: 30-Jun-2026
351.00
Save 52.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
298.65
Special offer
Online exclusive
Ends: 30-Jun-2026
351.00
Save 52.35 (15%)
Each
Chemical Identifiers
CAS14937-45-2
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline or fused solid, lumps or powder
Identification (FTIR)Conforms
Melting Point (clear melt)56-63°C
Appearance (Color)White to cream or pale yellow
Assay (Titration ex Bromide)≥98.0 to ≤102.0%
View more
(n-Hexadecyl)tri-n-butylphosphonium bromide is used as Phase-transfer-catalyst. It is a eco friendly highly effective water treatment agent with low foaming and toxic-free.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(n-Hexadecyl)tri-n-butylphosphonium bromide is used as Phase-transfer-catalyst. It is a eco friendly highly effective water treatment agent with low foaming and toxic-free.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Protect from carbondioxide. Store away from moisture.
RUO – Research Use Only