1-Bromo-2,3,5,6-tetramethylbenzene, 99%
1-Bromo-2,3,5,6-tetramethylbenzene, 99%
1-Bromo-2,3,5,6-tetramethylbenzene, 99%
Thermo Scientific Chemicals

1-Bromo-2,3,5,6-tetramethylbenzene, 99%

CAS: 1646-53-3 | C10H13Br | 213.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01465.14
also known as L01465-14
Price (USD)
224.65
Online Exclusive
249.00
Save 24.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
224.65
Online Exclusive
249.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS1646-53-3
IUPAC Name3-bromo-1,2,4,5-tetramethylbenzene
Molecular FormulaC10H13Br
InChI KeyWJKBPTLQJXKEHC-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C)C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (GC)≥98.5%
Melting Point (clear melt)57-64°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only