2-Carboxycinnamic acid, predominantly trans, 97%
2-Carboxycinnamic acid, predominantly trans, 97%
2-Carboxycinnamic acid, predominantly trans, 97%
Thermo Scientific Chemicals

2-Carboxycinnamic acid, predominantly trans, 97%

CAS: 612-40-8 | C10H8O4 | 192.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01467.14
also known as L01467-14
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
174.65
Online Exclusive
194.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS612-40-8
IUPAC Name2-[(1E)-2-carboxyeth-1-en-1-yl]benzoic acid
Molecular FormulaC10H8O4
InChI KeySCWPNMHQRGNQHH-AATRIKPKSA-N
SMILESOC(=O)\C=C\C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (HPLC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only