Thermo Scientific Chemicals

Norcamphor, 98+%, Thermo Scientific Chemicals

Catalog number: L01491.06
5 g, Each
Thermo Scientific Chemicals

Norcamphor, 98+%, Thermo Scientific Chemicals

Catalog number: L01491.06
5 g, Each
Quantity
Catalog number: L01491.06
also known as L01491-06
Price (USD)
Price: 40.30
Online price: 34.65
Your price:
Quantity
-

Chemical Identifiers

CAS
497-38-1
IUPAC Name
bicyclo[2.2.1]heptan-2-one
Molecular Formula
C7H10O
InChI Key
KPMKEVXVVHNIEY-UHFFFAOYNA-N
SMILES
O=C1CC2CCC1C2
Melting Point (clear melt)
86-99?C
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Appearance (Color)
Colorless to white to cream
Form
Waxy crystalline solid or compacted crystalline powder

Description

Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text