2-Amino-5-methylbenzoic acid, 97%
2-Amino-5-methylbenzoic acid, 97%
2-Amino-5-methylbenzoic acid, 97%
Thermo Scientific Chemicals

2-Amino-5-methylbenzoic acid, 97%

CAS: 2941-78-8 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01513.06
also known as L01513-06
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS2941-78-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to yellow to brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder or lumps or fused solid or granules
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only