2,4-Dichlorobenzeneboronic acid, 98%
2,4-Dichlorobenzeneboronic acid, 98%
2,4-Dichlorobenzeneboronic acid, 98%
Thermo Scientific Chemicals

2,4-Dichlorobenzeneboronic acid, 98%

CAS: 68716-47-2 | C6H5BCl2O2 | 190.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L01563.03
also known as L01563-03
Price (USD)
22.65
Special offer
Online exclusive
Ends: 30-Jun-2026
26.00
Save 3.35 (13%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
22.65
Special offer
Online exclusive
Ends: 30-Jun-2026
26.00
Save 3.35 (13%)
Each
Chemical Identifiers
CAS68716-47-2
IUPAC Name(2,4-dichlorophenyl)boronic acid
Molecular FormulaC6H5BCl2O2
InChI KeyQNEGDGPAXKYZHZ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5% (UK sourced material only)
FormCrystals or powder or crystalline powder or lumps or fused solid or granules
Assay (HPLC)≥97.5%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only