3-Methyl-2-furoic acid, 98+%
3-Methyl-2-furoic acid, 98+%
3-Methyl-2-furoic acid, 98+%
Thermo Scientific Chemicals

3-Methyl-2-furoic acid, 98+%

CAS: 4412-96-8 | C6H6O3 | 126.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L01763.03
also known as L01763-03
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.60
Save 11.95 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.60
Save 11.95 (14%)
Each
Chemical Identifiers
CAS4412-96-8
IUPAC Name3-methylfuran-2-carboxylic acid
Molecular FormulaC6H6O3
InChI KeyBNYIQEFWGSXIKQ-UHFFFAOYSA-N
SMILESCC1=C(OC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or granules
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only