1-Bromo-1-chloroethane, 98%
1-Bromo-1-chloroethane, 98%
1-Bromo-1-chloroethane, 98%
Thermo Scientific Chemicals

1-Bromo-1-chloroethane, 98%

CAS: 593-96-4 | C2H4BrCl | 143.408 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L01910.09
also known as L01910-09
Price (USD)
309.65
Special offer
Online exclusive
Ends: 30-Jun-2026
364.00
Save 54.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
309.65
Special offer
Online exclusive
Ends: 30-Jun-2026
364.00
Save 54.35 (15%)
Each
Chemical Identifiers
CAS593-96-4
IUPAC Name1-bromo-1-chloroethane
Molecular FormulaC2H4BrCl
InChI KeyQMSVNDSDEZTYAS-UHFFFAOYNA-N
SMILESCC(Cl)Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)>97.5%
Refractive Index1.4635-1.4685 @ 20?C
Appearance (Color)Clear colorless to pale yellow
1-Bromo-1-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-1-chloroethane is used as a solvent and as an intermediate for organic synthesis. It is used as a as fumigan.

Solubility
Difficult in mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. James H. Espenson.; Dennis A. Williams. Kinetics and mechanism of the cleavage reactions of alkylchromium cations with bromine. J. Am. Chem. Soc. 1974, 96, (4), 1008-1012.
  2. William J. DunnIII.; Silvio L. Emery.; W. Graham Glen.; Donald R. Scott. Preprocessing, variable selection, and classification rules in the application of SIMCA pattern recognition to mass-spectral data. Environ. Sci. Techno. 1989, 23, (12), 1499-1505.