2,5-Dimethylbenzaldehyde, 98%
2,5-Dimethylbenzaldehyde, 98%
2,5-Dimethylbenzaldehyde, 98%
Thermo Scientific Chemicals

2,5-Dimethylbenzaldehyde, 98%

CAS: 5779-94-2 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L01924.06
also known as L01924-06
Price (USD)
69.65
Online Exclusive
76.80
Save 7.15 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
76.80
Save 7.15 (9%)
Each
Chemical Identifiers
CAS5779-94-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Free acid (titration)≤1.5%
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
View more
2,5-Dimethylbenzaldehyde is useful for building asymmetric top molecules. 2,5-DMBA has also been used to analyze vehicle carbon emissions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylbenzaldehyde is useful for building asymmetric top molecules. 2,5-DMBA has also been used to analyze vehicle carbon emissions.

Solubility
Not miscible in water.

Notes
Air Sensitive. Store under dry inert gas. Do not expose to air. Incompatible with Air, strong bases, strong oxidizing agents.
RUO – Research Use Only

General References:

  1. A. Després; E. Migirdicyan. Determinations of large ZFS D-parameters in 2,4-, 2,5- and 3,4-dimethybenzaldehydes dispersed in durene single crystals.Chemical Physics.1977, 22 121-130.
  2. I. C. Winkler and David M. Hanson. Variation of dipole moments with triplet state energy spacings in substituted benzaldehydes.J. Chem. Phys.1983, 78 4836.