4,5-Dibromoveratrole, 98+%
4,5-Dibromoveratrole, 98+%
4,5-Dibromoveratrole, 98+%
4,5-Dibromoveratrole, 98+%
Thermo Scientific Chemicals

4,5-Dibromoveratrole, 98+%

CAS: 37895-73-1 | C8H8Br2O2 | 295.958 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number L01999.18
also known as L01999-18
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
158.65
Online Exclusive
176.00
Save 17.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS37895-73-1
IUPAC Name1,2-dibromo-4,5-dimethoxybenzene
Molecular FormulaC8H8Br2O2
InChI KeyZYCLQXMMFJREPJ-UHFFFAOYSA-N
SMILESCOC1=CC(Br)=C(Br)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.0%
Appearance (Color)White to pale cream to pale brown
Identification (FTIR)Conforms
4,5-Dibromoveratrole is used as chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dibromoveratrole is used as chemical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with Strong oxidizing agents.
RUO – Research Use Only