Tropinone, 99%
Tropinone, 99%
Tropinone, 99%
Thermo Scientific Chemicals

Tropinone, 99%

CAS: 532-24-1 | C8H13NO | 139.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L02011.06
also known as L02011-06
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
37.70
Save 5.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
37.70
Save 5.05
Each
Chemical Identifiers
CAS532-24-1
IUPAC Name8-methyl-8-azabicyclo[3.2.1]octan-3-one
Molecular FormulaC8H13NO
InChI KeyQQXLDOJGLXJCSE-UHFFFAOYNA-N
SMILESCN1C2CCC1CC(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow or brown
FormCrystal or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)38-45?C
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only