3-Methoxybenzylamine, 97%
3-Methoxybenzylamine, 97%
3-Methoxybenzylamine, 97%
Thermo Scientific Chemicals

3-Methoxybenzylamine, 97%

CAS: 5071-96-5 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02054.06
also known as L02054-06
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.90
Save 8.25 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.90
Save 8.25 (14%)
Each
Chemical Identifiers
CAS5071-96-5
IUPAC Name1-(3-methoxyphenyl)methanamine
Molecular FormulaC8H11NO
InChI KeyGRRIMVWABNHKBX-UHFFFAOYSA-N
SMILESCOC1=CC=CC(CN)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5455-1.5495 @ 20°C
FormLiquid
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Assay (GC)≥96.0%
3-Methoxybenzylamine is used in the preparation of 6-substituted purines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzylamine is used in the preparation of 6-substituted purines

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
A. Torres-Gavilan.; E. Castillo.; A. Lopez-Munguia. The amidase activity of Candida antarctica lipase B is dependent on specific structural features of the substrates. Journal of Molecular Catalysis B: Enzymatic. 2006, 41, (3-4), 136-140.