2-Aminobenzimidazole, 97+%
2-Aminobenzimidazole, 97+%
2-Aminobenzimidazole, 97+%
Thermo Scientific Chemicals

2-Aminobenzimidazole, 97+%

CAS: 934-32-7 | C7H7N3 | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L02066.06
also known as L02066-06
Price (USD)
33.65
Online Exclusive
36.80
Save 3.15 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
36.80
Save 3.15 (9%)
Each
Chemical Identifiers
CAS934-32-7
IUPAC Name1H-1,3-benzodiazol-2-amine
Molecular FormulaC7H7N3
InChI KeyJWYUFVNJZUSCSM-UHFFFAOYSA-N
SMILESNC1=NC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
FormCrystalline powder or crystals and/or flakes
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0%%
Assay (HPLC)≥97.0%
CommentMay darken on storage
View more
2-Aminobenzimidazole is used as receptors in chemosensor analysis. It was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzimidazole is used as receptors in chemosensor analysis. It was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.

Solubility
Soluble in water (Slightly).

Notes
Protect from direct light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Jongmin Kang; Hyung Suk Kima; Doo Ok Jang. Fluorescent anion chemosensors using 2-aminobenzimidazole receptors.Tetrahedron Letters.2005, 46 6079-6082.
  2. Wanda Nawrocka; Barbara Sztuba; Maria W Kowalska; Hanna Liszkiewicz; Joanna Wietrzyk; Anna Nasulewicz; Marzena Pełczyńska; Adam Opolski. Synthesis and antiproliferative activity in vitro of 2-aminobenzimidazole derivatives.Il Farmaco.2004, 59 83-91.
  3. For a review of the use of 2-aminobenzimidazoles in organic synthesis, see: Synthesis, 861 (1983).