p-Toluenesulfonylacetic acid, 98%
p-Toluenesulfonylacetic acid, 98%
p-Toluenesulfonylacetic acid, 98%
Thermo Scientific Chemicals

p-Toluenesulfonylacetic acid, 98%

CAS: 3937-96-0 | C9H10O4S | 214.235 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02102.14
also known as L02102-14
Price (USD)
378.65
Online Exclusive
420.00
Save 41.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
378.65
Online Exclusive
420.00
Save 41.35 (10%)
Each
Specifications
Beilstein2331311
CAS3937-96-0
Chemical Name or Materialp-Toluenesulfonylacetic acid
EINECS Number223-518-0
Formula Weight214.24
View more
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only