4'-Hydroxynonanophenone, 96%
4'-Hydroxynonanophenone, 96%
4'-Hydroxynonanophenone, 96%
Thermo Scientific Chemicals

4'-Hydroxynonanophenone, 96%

CAS: 14392-69-9 | C15H22O2 | 234.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02293.06
also known as L02293-06
Price (USD)
66.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.00
Each
Chemical Identifiers
CAS14392-69-9
IUPAC Name1-(4-hydroxyphenyl)nonan-1-one
Molecular FormulaC15H22O2
InChI KeyJZBXYOOARNRUME-UHFFFAOYSA-N
SMILESCCCCCCCCC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Melting Point (clear melt)51.0-60.0?C
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder or lumps or fused solid or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only