Diphenyl-n-propylphosphine, 97%
Diphenyl-n-propylphosphine, 97%
Diphenyl-n-propylphosphine, 97%
Thermo Scientific Chemicals

Diphenyl-n-propylphosphine, 97%

CAS: 7650-84-2 | C15H17P | 228.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L02402.03
also known as L02402-03
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.70
Save 6.05
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.70
Save 6.05
Each
Chemical Identifiers
CAS7650-84-2
IUPAC Namediphenyl(propyl)phosphane
Molecular FormulaC15H17P
InChI KeyAAXGWYDSLJUQLN-UHFFFAOYSA-N
SMILESCCCP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Phosphorus-31 NMRConforms to structure
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay from Suppliers CofA≥96.0% (GC)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only