(1-Octyl)triphenylphosphonium bromide, 97%
(1-Octyl)triphenylphosphonium bromide, 97%
(1-Octyl)triphenylphosphonium bromide, 97%
Thermo Scientific Chemicals

(1-Octyl)triphenylphosphonium bromide, 97%

CAS: 42036-78-2 | C26H32BrP | 455.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L02412.06
also known as L02412-06
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Chemical Identifiers
CAS42036-78-2
IUPAC Nameoctyltriphenylphosphanium bromide
Molecular FormulaC26H32BrP
InChI KeyOBLXVLWZBMAMHE-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or lumps
Assay (Titration ex Bromide)≥96.0 to ≤104.0%
Melting Point (clear melt)58.0-67.0?C
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
RUO – Research Use Only