4-tert-Butylcyclohexylamine, cis + trans, 97%
4-tert-Butylcyclohexylamine, cis + trans, 97%
4-tert-Butylcyclohexylamine, cis + trans, 97%
Thermo Scientific Chemicals

4-tert-Butylcyclohexylamine, cis + trans, 97%

CAS: 5400-88-4 | C10H21N | 155.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02472.06
also known as L02472-06
Price (USD)
50.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
50.20
Each
Chemical Identifiers
CAS5400-88-4
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4630-1.4680 @ 20?C
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0% (sum of isomers)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only