1-Tridecyne, 97%
1-Tridecyne, 97%
1-Tridecyne, 97%
Thermo Scientific Chemicals

1-Tridecyne, 97%

CAS: 26186-02-7 | C13H24 | 180.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L02481.06
also known as L02481-06
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Specifications
Beilstein1744658
CAS26186-02-7
Chemical Name or Material1-Tridecyne
Density0.78
EINECS Number247-511-7
View more
It finds its application in the synthesis of unsaturated fatty acids and in the stereoselective synthesis of d-erythro- and l-threo-sphinganines via palladium-catalyzed equilibration and Suzuki coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of unsaturated fatty acids and in the stereoselective synthesis of d-erythro- and l-threo-sphinganines via palladium-catalyzed equilibration and Suzuki coupling.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
RUO – Research Use Only

General References:

  1. Nico Klewer, et al. (Z)-Pentacos-12-ene, an Oviposition-deterring Pheromone of Cheilomenes sexmaculata.J. Chem. Ecol.,2007,33(11), 2167-2170.
  2. Ian N.H. White, et al. Structure-activity relationships in the destruction of cytochrome P-450 mediated by certain ethynyl-substituted compounds in rats.Biochem. Pharmacol.,1980,29(24), 3253-3255.