2,3,5-Triiodobenzoic acid, 98+%
2,3,5-Triiodobenzoic acid, 98+%
2,3,5-Triiodobenzoic acid, 98+%
Thermo Scientific Chemicals

2,3,5-Triiodobenzoic acid, 98+%

CAS: 88-82-4 | C7H3I3O2 | 499.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L02679.06
also known as L02679-06
Price (USD)
53.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.80
Each
Chemical Identifiers
CAS88-82-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to to cream to pale brown
Assay from Supplier's CofA≥98.0%
Melting Point218-221°C
FormPowder
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Triiodobenzoic acid has been used in the synthesis of aromatic iodine containing vinylic monomer, 2-methacryloyloxyethyl(2,3,5-triiodobenzoate) used in preparing radiopaque polymers.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with oxidizing agents and light.
RUO – Research Use Only

General References:

  1. Anna Galperin; Shlomo Margel. Synthesis and characterization of radiopaque magnetic core-shell nanoparticles for X-ray imaging applications. Journal of Biomedical Materials Research, Part B: Applied Biomaterials. 2007, 83,(2), 490-498.
  2. J R Hay. The Effect of 2,4-Dichlorophenoxyacetic Acid and 2,3,5-Triiodobenzoic Acid on the Transport of Indoleacetic Acid. Plant Physiology.1956, 31,(2), 118-120.