1,7-Diaminoheptane, 98%
1,7-Diaminoheptane, 98%
1,7-Diaminoheptane, 98%
Thermo Scientific Chemicals

1,7-Diaminoheptane, 98%

CAS: 646-19-5 | C7H18N2 | 130.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02853.06
also known as L02853-06
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.70
Save 6.05 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.70
Save 6.05 (14%)
Each
Chemical Identifiers
CAS646-19-5
IUPAC Nameheptane-1,7-diamine
Molecular FormulaC7H18N2
InChI KeyPWSKHLMYTZNYKO-UHFFFAOYSA-N
SMILESNCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Colorless to white to pale cream to cream to yellow
FormCrystals or powder or crystalline powder or lumps or fused solid or clear liquid as melt
Identification (FTIR)Conforms
Melting Point (clear melt)24.0-31.0?C
View more
1,7-Diaminoheptane is used in the preparation of 7-amino-1-guanidinooctane and 1,7-diamino-trans-hept-3-ene, which is used as a deoxyhypusine synthase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,7-Diaminoheptane is used in the preparation of 7-amino-1-guanidinooctane and 1,7-diamino-trans-hept-3-ene, which is used as a deoxyhypusine synthase inhibitor.

Solubility
Soluble in water, ethanol, acetone, ether and benzene.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Doria, F.; Oppi, A.; Manoli, F.; Botti, S.; Kandoth, N.; Grande, V.; Manet, I.; Freccero, M. A naphthalene diimide dyad for fluorescence switch-on detection of G-quadruplexes. Chem. Commun. 2015, 51, 9105-9108.
  2. Daniel, D.; dos Santos, V. B.; Vidal, D. T. R.; do Lago, C. L. Determination of biogenic amines in beer and wine by capillary electrophoresis-tandem mass spectrometry. J. Chromatogr. A 2015, 1416, 121-128.