n-Butylurea, 96%
n-Butylurea, 96%
n-Butylurea, 96%
Thermo Scientific Chemicals

n-Butylurea, 96%

CAS: 592-31-4 | C5H12N2O | 116.164 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L02854.36
also known as L02854-36
Price (USD)
633.65
Online Exclusive
704.00
Save 70.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
633.65
Online Exclusive
704.00
Save 70.35 (10%)
Each
Chemical Identifiers
CAS592-31-4
IUPAC Namebutylurea
Molecular FormulaC5H12N2O
InChI KeyCNWSQCLBDWYLAN-UHFFFAOYSA-N
SMILESCCCCNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)≥95.0 to ≤105.0% (based on elemetal N, C)
Elemental AnalysisC: 49.12-54.29% (Theory: 51.7%)
Elemental AnalysisN: 22.91-25.32% (Theory: 24.12%)
Water Content (Karl Fischer Titration)≤2%
Melting Point (clear melt)88.0-98.0?C
View more
n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Butylurea is used as a fertilizer for rice. It is used in the preparation of drugs which are recommended for diabetes.

Solubility
Soluble in water, Alcohol, Ether.

Notes
Incompatible with Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
RUO – Research Use Only

General References:

  1. L. Paduano; R. Sartorio; V. Vitagliano; L. Costantino. Diffusion coefficients of the system α-cyclodextrin-n-butylurea-water at 25°C.Journal of Solution Chemistry.1995, 24 1143-1153.
  2. Monika Obrzud; Maria Rospenk and Aleksander Kolla. Self-aggregation mechanisms of N-alkyl derivatives of urea and thiourea.Phys. Chem. Chem. Phys.2014, 16 3209-3219.