Methyl p-tolyl sulfide, 97%
Methyl p-tolyl sulfide, 97%
Methyl p-tolyl sulfide, 97%
Methyl p-tolyl sulfide, 97%
Thermo Scientific Chemicals

Methyl p-tolyl sulfide, 97%

CAS: 623-13-2 | C8H10S | 138.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02933.14
also known as L02933-14
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS623-13-2
IUPAC Name1-methyl-4-(methylsulfanyl)benzene
Molecular FormulaC8H10S
InChI KeyVHILIAIEEYLJNA-UHFFFAOYSA-N
SMILESCSC1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.5710-1.5760 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only