1-Dodecyne, 97%
1-Dodecyne, 97%
1-Dodecyne, 97%
Thermo Scientific Chemicals

1-Dodecyne, 97%

CAS: 765-03-7 | C12H22 | 166.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L02977.06
also known as L02977-06
Price (USD)
48.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
48.00
Each
Chemical Identifiers
CAS765-03-7
IUPAC Namedodec-1-yne
Molecular FormulaC12H22
InChI KeyZVDBUOGYYYNMQI-UHFFFAOYSA-N
SMILESCCCCCCCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4320-1.4370 @ 20°C (non-U.S. specification)
FormLiquid
1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

Solubility
Not miscible in water.

Notes
Incompatible with heavy metal powders and oxidizing agents. Store between 2 - 8°C. Store away from oxidizing agents.
RUO – Research Use Only
Xiongwu Kang; Nathaniel B Zuckerman; Joseph P Konopelski; Shaowei Chen. Alkyne-functionalized ruthenium nanoparticles: ruthenium-vinylidene bonds at the metal-ligand interface.Journal of the American Chemical Society.2012, 134 (3), 1412-1415.