p-Xylylenediamine is
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Xylylenediamine is
Solubility
Soluble in water.
Notes
Air Sensitive. Incompatible with air, carbondioxide and oxidizing agents. Store underdry inert gas.
RUO – Research Use Only
Kazuhiko Hashimoto; Masahiko Okada and Naomi Honjou. Ring-opening polyaddition of D-glucaro-1,4:6,3-dilactone with p-xylylenediamine.Molecular Rapid Communications.2003, 11 393-396.