p-Xylylenediamine, 97%
p-Xylylenediamine, 97%
p-Xylylenediamine, 97%
Thermo Scientific Chemicals

p-Xylylenediamine, 97%

CAS: 539-48-0 | C8H12N2 | 136.198 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number L02995.18
also known as L02995-18
Price (USD)
374.65
Special offer
Online exclusive
Ends: 30-Jun-2026
441.00
Save 66.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
374.65
Special offer
Online exclusive
Ends: 30-Jun-2026
441.00
Save 66.35 (15%)
Each
Chemical Identifiers
CAS539-48-0
IUPAC Name1-[4-(aminomethyl)phenyl]methanamine
Molecular FormulaC8H12N2
InChI KeyISKQADXMHQSTHK-UHFFFAOYSA-N
SMILESNCC1=CC=C(CN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Melting Point (clear melt)57.0-65.0°C
p-Xylylenediamine is

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylenediamine is

Solubility
Soluble in water.

Notes
Air Sensitive. Incompatible with air, carbondioxide and oxidizing agents. Store underdry inert gas.
RUO – Research Use Only
Kazuhiko Hashimoto; Masahiko Okada and Naomi Honjou. Ring-opening polyaddition of D-glucaro-1,4:6,3-dilactone with p-xylylenediamine.Molecular Rapid Communications.2003, 11 393-396.