3-Methylpentane, 99+%
3-Methylpentane, 99+%
3-Methylpentane, 99+%
Thermo Scientific Chemicals

3-Methylpentane, 99+%

CAS: 96-14-0 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L03133.AP
also known as L03133-AP
Price (USD)
176.65
Special offer
Online exclusive
Ends: 30-Jun-2026
208.00
Save 31.35 (15%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
176.65
Special offer
Online exclusive
Ends: 30-Jun-2026
208.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS96-14-0
IUPAC Name3-methylpentane
Molecular FormulaC6H14
InChI KeyPFEOZHBOMNWTJB-UHFFFAOYSA-N
SMILESCCC(C)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥99.0%
Identification (FTIR)Conforms
Refractive Index1.3755-1.3785 @ 20?C
FormLiquid
3-Methylpentane is used as a solvent in the preparation of vegetable oils, glues, coatings, and paints. It is also used in gasoline, rubber solvent, and petroleum ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methylpentane is used as a solvent in the preparation of vegetable oils, glues, coatings, and paints. It is also used in gasoline, rubber solvent, and petroleum ether.

Solubility
Soluble in water. 0.013 g/L at 20°C). Miscible in ether, acetone. Soluble in ethanol.

Notes
Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only

General References:

  1. Stein Arnold; Allegra Joseph C; Allen Guy F. Shear Viscosity of 3-Methylpentane—Nitroethane near the Critical Point.The Journal of Chemical Physics.1971, 55 4265-4269.
  2. Milton Burton; Michael Dillon and Robert Rein. Luminescence Phenomena in Gamma-Irradiated Pure 3-Methylpentane Glass: Implications for Radiation Chemistry.J. Chem. Phys.1964, 41 2228.