3-Methyl-4-nitrophenol, 98%
3-Methyl-4-nitrophenol, 98%
3-Methyl-4-nitrophenol, 98%
3-Methyl-4-nitrophenol, 98%
Thermo Scientific Chemicals

3-Methyl-4-nitrophenol, 98%

CAS: 2581-34-2 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L03163.22
also known as L03163-22
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
93.65
Special offer
Online exclusive
Ends: 30-Jun-2026
110.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS2581-34-2
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5% (HPLC, U.S. sourced material)
Melting Point (clear melt)125.0-131.0°C (non-U.S. sourced material)
FormCrystals or powder or crystalline powder
Appearance (Color)Cream to brown to yellow
Assay (GC)≥97.5% (non-U.S. sourced material)
View more
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

Solubility
Soluble in water(1.34g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chunmei Li,; Shinji Taned,; Akira K. Suzuki,;Chie Furuta. Effects of 3-Methyl-4-Nitrophenol in Diesel Exhaust Particles on the Regulation of Testicular Function in Immature Male Rats. Journal of Andrology. 2007, 28(2), 252-258.
  2. ChunMei Li,; Shinji Taned,; Akira K. Suzuk. Anti-androgenic activity of 3-methyl-4-nitrophenol in diesel exhaust particles. European Journal of Pharmacology. 2009, 543 (1-3) , 194-199.