Thermo Scientific Chemicals

Methoxyacetyl chloride, 97%, stab. with ca 0.3% magnesium oxide, Thermo Scientific Chemicals

Catalog number: L03210.14
25 g, Each
Thermo Scientific Chemicals

Methoxyacetyl chloride, 97%, stab. with ca 0.3% magnesium oxide, Thermo Scientific Chemicals

Catalog number: L03210.14
25 g, Each
Quantity
Catalog number: L03210.14
also known as L03210-14
Price (USD)
Price: 154.00
Online price: 130.65
Your price:
Quantity
-

Chemical Identifiers

CAS
38870-89-2
IUPAC Name
2-methoxyacetyl chloride
Molecular Formula
C3H5ClO2
InChI Key
JJKWHOSQTYYFAE-UHFFFAOYSA-N
SMILES
COCC(Cl)=O
Refractive Index
1.4190 - 1.4290 @20?C
Assay (GC)
> 96.0%
Form
liquid with slight sediment
Appearance (Color)
Colourless to pale yellow

Description

Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetyl chloride is used as an intermediate for active pharmaceutical ingredients and dyes. Further, it acts as a precursor for the synthesis of agrochemicals.

Solubility
Miscible with acetone, ether, carbon tetrachloride and chloroform.

Notes
Moisture sensitive. Store in a cool place. Incompatible with water, alcohols, oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text