Ethyl 2-methylnicotinate, 97%
Ethyl 2-methylnicotinate, 97%
Ethyl 2-methylnicotinate, 97%
Thermo Scientific Chemicals

Ethyl 2-methylnicotinate, 97%

CAS: 1721-26-2 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L03256.06
also known as L03256-06
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.90
Save 10.25 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.90
Save 10.25 (15%)
Each
Chemical Identifiers
CAS1721-26-2
IUPAC Nameethyl 2-methylpyridine-3-carboxylate
Molecular FormulaC9H11NO2
InChI KeyTZORNSWZUZDUCU-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CN=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to dark yellow to orange or yellow-orange
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5020-1.5080 @ 20°C
Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. K. Tsuda; Y. Satch; N. Ikekawa; H. Mishima. Notes - Reaction Between Acrolein and Ethyl β-Aminocrotonate. J. Org. Chem. 1956, 21, (7), 800-801.
  2. J. Hurst and D. G. Wibberley. 2-Bromomethylnicotinates: reaction with nucleophilic reagents. J. Chem. Soc. 1962, 119-122.