Thioacetic acid, 97%
Thioacetic acid, 97%
Thioacetic acid, 97%
Thermo Scientific Chemicals

Thioacetic acid, 97%

CAS: 507-09-5 | C2H4OS | 76.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L03305.14
also known as L03305-14
Price (USD)
32.65
Online Exclusive
35.90
Save 3.25 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
35.90
Save 3.25 (9%)
Each
Chemical Identifiers
CAS507-09-5
IUPAC Nameethanethioic S-acid
Molecular FormulaC2H4OS
InChI KeyDUYAAUVXQSMXQP-UHFFFAOYSA-N
SMILESCC(S)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4610-1.4665 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to yellow
Assay (Argentometric Titration)≥96.0%
Identification (FTIR)Conforms

Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).

Solubility
Soluble in water. (27g/L) at 16°C.

Notes
Protect from heat. Store away from oxidizing agents. Incompatible with strong bases, strong oxidizing agents, metals.
RUO – Research Use Only

General References:

  1. Wen Yang; Da-Ming Du. Squaramide-catalysed enantio- and diastereoselective sulfa-Michael addition of thioacetic acid to α,β-disubstituted nitroalkenes.Organic & Biomolecular Chemistry.2012, 10 (34), 6876-6884.
  2. Libo Hu; Hui Zhu; Da-Ming Du; Jiaxi Xu. Efficient synthesis of taurine and structurally diverse substituted taurines from aziridines.Journal of Organic Chemistry.2007, 72 (12), 4543-4546.
  3. Used in several different ways for the introduction of the thiol group: Radical addition to unactivated alkenes: J. Chem. Soc., 2123 (1951). Conjugate (Michael) addition to activated double bonds: J. Am. Chem. Soc., 77, 5144 (1955); 81, 1224 (1959). Nucleophilic displacement of halide, tosylate or acetate: J. Am. Chem. Soc., 73, 2659 (1951); Carbohydr. Res., 17, 457 (1971). Alcohols can be converted to thiol esters under Mitsunobu conditions, followed by LAH reduction to the thiol: Tetrahedron Lett., 3119 (1981).