4-Methylsalicylic acid, 99%
4-Methylsalicylic acid, 99%
4-Methylsalicylic acid, 99%
4-Methylsalicylic acid, 99%
Thermo Scientific Chemicals

4-Methylsalicylic acid, 99%

CAS: 50-85-1 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L03367.22
also known as L03367-22
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS50-85-1
IUPAC Name2-hydroxy-4-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyNJESAXZANHETJV-UHFFFAOYSA-N
SMILESCC1=CC=C(C(O)=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale orange to gray
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylsalicylic acid was used to inhibit the medium chain acyl- CoA synthetase.

Solubility
Soluble in water 10 g/L @ 20˚C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. F Kasuya; K Igarashi; M Fukui. Inhibition of a medium chain acyl-CoA synthetase involved in glycine conjugation by carboxylic acids. Biochemical Pharmacology. 1996, 52, (10), 1643-1646.
  2. Xiao-xia Guo; Yu-ying Hao; Jun-feng Lei; Wen-hao Fan; Hua Wang; Bing-she Xu. [New highly phosphorescent heteroleptic tris-cyclometalated iridium (III) complexes: synthesis and photophysical characterization]. Guang pu xue yu guang pu fen xi = Guang pu. 2010, 30, (1), 170-173.