4-(Bromomethyl)benzoic acid, 97%
4-(Bromomethyl)benzoic acid, 97%
4-(Bromomethyl)benzoic acid, 97%
4-(Bromomethyl)benzoic acid, 97%
Thermo Scientific Chemicals

4-(Bromomethyl)benzoic acid, 97%

CAS: 6232-88-8 | C8H7BrO2 | 215.046 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L03371.14
also known as L03371-14
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS6232-88-8
IUPAC Name4-(bromomethyl)benzoic acid
Molecular FormulaC8H7BrO2
InChI KeyCQQSQBRPAJSTFB-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale grey
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
Identification (FTIR)Conforms
4-(Bromomethyl)benzoic acid acts as an intermediate in the synthesis of eprosartan, which is used as an antihypertensive agent. Further, it is used in the preparation of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), which is a second generation photosensitizer. In addition to this, it is used in the preparation of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Bromomethyl)benzoic acid acts as an intermediate in the synthesis of eprosartan, which is used as an antihypertensive agent. Further, it is used in the preparation of 5,10,15,20-tetra(m-hydroxyphenyl)chlorin (temoporfin), which is a second generation photosensitizer. In addition to this, it is used in the preparation of 4-(5-arylidene-2,4-dioxothiazolidin-3-yl) methylbenzoic acids.

Solubility
Soluble in water.

Notes
Incompatible with bases, amines and oxidizing agents.
RUO – Research Use Only

General References:

  1. Eyley, S.; Vandamme, D.; Lama, S.; Van den Mooter, G.; Muylaert, K.; Thielemans, W. CO2 controlled flocculation of microalgae using pH responsive cellulose nanocrystals. Nanoscale 2015, 7 (34), 14413-14421.
  2. Paraskevopoulos, G.; Alivertis, D.; Theodorou, V.; Skobridis, K. New Calix[4]arene-Cored Peripherally Functionalized Dendrimers: Synthesis and Conformational Characteristics. Helv. Chim. Acta 2015, 98 (5), 593-603.