2-Bromo-1,3-diethylbenzene, 94%
2-Bromo-1,3-diethylbenzene, 94%
2-Bromo-1,3-diethylbenzene, 94%
2-Bromo-1,3-diethylbenzene, 94%
Thermo Scientific Chemicals

2-Bromo-1,3-diethylbenzene, 94%

CAS: 65232-57-7 | C10H13Br | 213.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L03482.03
also known as L03482-03
Price (USD)
59.65
Online Exclusive
65.80
Save 6.15 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
59.65
Online Exclusive
65.80
Save 6.15 (9%)
Each
Add to cart
Chemical Identifiers
CAS65232-57-7
IUPAC Name2-bromo-1,3-diethylbenzene
Molecular FormulaC10H13Br
InChI KeyFPWGIABDOFXETH-UHFFFAOYSA-N
SMILESCCC1=CC=CC(CC)=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange to red
Assay (GC)≥92.0%
Refractive Index1.5380-1.5475 @ 20?C
FormLiquid
2-Bromo-1,3-diethylbenzene is used as raw material in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,3-diethylbenzene is used as raw material in synthetic organic chemistry.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ding, C.; Fan, C.; Jiang, G.; Zhang, J.; Li, X.; Li, N.; Pan, X.; Zhang, Z.; Zhang, W.; Zhu, J.; Zhu, X. Diselenide mediated controlled radical polymerization under visible light irradiation: mechanism investigation and alpha,μ-ditelechelic polymers. Polym. Chem. 2015, 6 (35), 6416-6423.
  2. Egan, B. A.; Burton, P. M. Synthesis of 3-aryl-1H-indazoles via iridium-catalysed C-H borylation and Suzuki-Miyaura coupling. RSC Adv. 2014, 4 (53), 27726-27729.