2-Methyl-1-phenyl-2-propanol, 98+%
2-Methyl-1-phenyl-2-propanol, 98+%
2-Methyl-1-phenyl-2-propanol, 98+%
Thermo Scientific Chemicals

2-Methyl-1-phenyl-2-propanol, 98+%

CAS: 100-86-7 | C10H14O | 150.221 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L03679.36
also known as L03679-36
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS100-86-7
IUPAC Name2-methyl-1-phenylpropan-2-ol
Molecular FormulaC10H14O
InChI KeyRIWRBSMFKVOJMN-UHFFFAOYSA-N
SMILESCC(C)(O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale yellow
FormCrystals or powder or crystalline powder or fused solid or clear liquid or viscous liquid as melt
Assay (GC)≥98.0%
Refractive Index1.5135-1.5175 @ 20?C
Identification (FTIR)Conforms
2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Takao Raku; Yutaka Tokiwa. Chemoenzymatic synthesis of optically active, biodegradable polymers based on phenyl- and naphthyl-ethanols esterified with divinyladipate. Biotechnology Letters. 2004, 26, (8), 665-670.
  2. David C. Magria and Mark S. Workentin. Model dialkyl peroxides of the Fenton mechanistic probe 2-methyl-1-phenyl-2-propyl hydroperoxide (MPPH): kinetic probes for dissociative electron transfer. Org. Biomol. Chem. 2003, 1, (19), 3418-3429.