2,3-Lutidine, 99%
2,3-Lutidine, 99%
2,3-Lutidine, 99%
Thermo Scientific Chemicals

2,3-Lutidine, 99%

CAS: 583-61-9 | C7H9N | 107.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L03697.36
also known as L03697-36
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS583-61-9
IUPAC Name2,3-dimethylpyridine
Molecular FormulaC7H9N
InChI KeyHPYNZHMRTTWQTB-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)>98.5%
FormLiquid
Refractive Index1.5065-1.5095 @ 20?C
2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.

Solubility
Soluble in water 95 gl/ @ 20˚C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Stanley Osman; J. Barson. The volatile bases of cigar smoke. Phytochemistry. 2004, 3, (5), 587-590.
  2. Harry H. Sisler; Wan Chen Loh Ming; Elaine Metter; Forrest R. Hurley. Addition Compounds of Chromium(VI) Oxide with Heterocyclic Nitrogen Bases. II. Reactions with the Lutidines, Vinylpyridines, 2,4,6-Collidine and α-Picoline. J. Am. Chem. Soc. 1953, 75, (2), 446-448.