Cyclohexyl methyl sulfide, 97%
Cyclohexyl methyl sulfide, 97%
Cyclohexyl methyl sulfide, 97%
Thermo Scientific Chemicals

Cyclohexyl methyl sulfide, 97%

CAS: 7133-37-1 | C7H14S | 130.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L03712.06
also known as L03712-06
Price (USD)
34.65
Online exclusive
38.30
Save 3.65
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
34.65
Online exclusive
38.30
Save 3.65
Each
Chemical Identifiers
CAS7133-37-1
IUPAC Name(methylsulfanyl)cyclohexane
Molecular FormulaC7H14S
InChI KeyQQBIOCGHCKNYGP-UHFFFAOYSA-N
SMILESCSC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4930-1.4980 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

Solubility
Soluble in water 173.5 mg/L @ 25°C, alcohol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Stefano Colonna; Nicoletta Gaggero; Giacomo Carrea and Piero Pasta. A new enzymatic enantioselective synthesis of dialkyl sulfoxidescatalysed by monooxygenases. Chemical Communications. 1997, (5), 439-440.