tert-Pentylbenzene, 97%
tert-Pentylbenzene, 97%
tert-Pentylbenzene, 97%
Thermo Scientific Chemicals

tert-Pentylbenzene, 97%

CAS: 2049-95-8 | C11H16 | 148.249 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L03731.18
also known as L03731-18
Price (USD)
155.65
Online Exclusive
173.00
Save 17.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
155.65
Online Exclusive
173.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS2049-95-8
IUPAC Name(2-methylbutan-2-yl)benzene
Molecular FormulaC11H16
InChI KeyQHTJSSMHBLGUHV-UHFFFAOYSA-N
SMILESCCC(C)(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
CommentMaterial sourced in the U.S. and in other countries
Assay (GC)≥96.0%
Refractive Index1.4930-1.4980 @ 20°C (non-U.S. sourced material)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only