beta-Estradiol, 99% (dry wt.), ca 3% water
beta-Estradiol, 99% (dry wt.), ca 3% water
beta-Estradiol, 99% (dry wt.), ca 3% water
Thermo Scientific Chemicals

beta-Estradiol, 99% (dry wt.), ca 3% water

CAS: 50-28-2 | C18H24O2 | 272.388 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L03801.03
also known as L03801-03
Price (USD)
45.65
Online Exclusive
50.50
Save 4.85 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.50
Save 4.85 (10%)
Each
Chemical Identifiers
CAS50-28-2
IUPAC Name(1S,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Molecular FormulaC18H24O2
InChI KeyVOXZDWNPVJITMN-ZBRFXRBCSA-N
SMILESC[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (Silylated GC)> 98.5%
Water Content (Karl Fischer Titration)<4.0%
Optical Rotation+79.0? ? 3.0? (c=1 in Dioxane)
β-Estradiol is the major estrogen secreted by the premenopausal ovary. It acts as an estrogenic hormone receptor (ER) agonist and activator of PI 3-kinase. Further, it is used in the treatment of infertility in women, prostate cancer and breast cancer. It is also employed as an agent of stimulating female puberty induction, as in the treatment of delayed puberty caused by hypogonadism or Turner's syndrome. In addition to this, it is used as part of the hormone replacement therapy for transgender women.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Estradiol is the major estrogen secreted by the premenopausal ovary. It acts as an estrogenic hormone receptor (ER) agonist and activator of PI 3-kinase. Further, it is used in the treatment of infertility in women, prostate cancer and breast cancer. It is also employed as an agent of stimulating female puberty induction, as in the treatment of delayed puberty caused by hypogonadism or Turner′s syndrome. In addition to this, it is used as part of the hormone replacement therapy for transgender women.

Solubility
Soluble in dimethyl sulfoxide, ethanol , water, phosphate buffer saline, dimethyl formamide, acetone, dioxane and alkali hydroxides. Slightly soluble in vegetable oils.

Notes
Incompatible with strong oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Liu, X.; Liu, Y.; Cheng, M.; Zhang, X.; Xiao, H. A metabolomics study of the inhibitory effect of 17-beta-estradiol on osteoclast proliferation and differentiation. Mol. Biosyst. 2015, 11 (2), 635-646.
  2. Pooley, A. E.; Luong, M.; Hussain, A.; Nathan, B. P. Neurite outgrowth promoting effect of 17-beta estradiol is mediated through estrogen receptor alpha in an olfactory epithelium culture. Brain Res. 2015, 1624, 19-27.