2-Ethylhexyl acrylate, 98%, stab.
2-Ethylhexyl acrylate, 98%, stab.
2-Ethylhexyl acrylate, 98%, stab.
Thermo Scientific Chemicals

2-Ethylhexyl acrylate, 98%, stab.

CAS: 103-11-7 | C11H20O2 | 184.279 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number L03854.AP
also known as L03854-AP
Price (USD)
49.65
Online Exclusive
54.80
Save 5.15 (9%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.80
Save 5.15 (9%)
Each
Chemical Identifiers
CAS103-11-7
IUPAC Name2-ethylhexyl prop-2-enoate
Molecular FormulaC11H20O2
InChI KeyGOXQRTZXKQZDDN-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Solution Test(10% v/v in diethyl ether) : Clear
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4335-1.4385 @ 20?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Ethylhexyl acrylate is used in the preparation of homopolymers and copolymers. It is involved in the copolymerization with acrylamide, methacrylic acid, acrylonitrile, styrene, maleic acid esters, vinyl acetate, vinyl chloride and unsaturated polyesters. Further, it acts as a starting material in chemical syntheses.

Solubility
Miscible with ether and alcohol. Immiscible with water.

Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Zhang, Y.; Dubé, M. A. Copolymerization of 2-Ethylhexyl Acrylate and D-Limonene. Polym. Plast. Technol. Eng. 2015, 54 (5), 499-505.
  2. Kockler, K. B.; Haehnel, A. P.; Fleischhaker, F.; Schneider-Baumann, M.; Misske, A. M.; Barner-Kowollik, C. No Apparent Correlation of kp with Steric Hindrance for Branched Acrylates. Macromol. Chem. Phys. 2015, 216 (14), 1573-1582.